EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences
Open-tubular CEC with a new triethanolamine bonded stationary phase for biomolecule separation.
PMID 25486613
Abstract
This study describes the preparation and electrochromatographic application of a new open-tubular capillary column with triethanolamine functionalized stationary phase. The stationary phase was synthesized by in situ grafting polymerization with 3-chloro-2-hydroxypropylmethacrylate based reactive monomer, and followed triethanolamine functionalization. The different 3-chloro-2-hydroxypropylmethacrylate contents on the separation efficiency were studied. The results indicated that the 3-chloro-2-hydroxypropylmethacrylate content (e.g., 15.3v/v%) on the inner surface of the capillary was very important for final preparation of the polymer stationary phase. The electrochromatographic characterization of the stationary phase was performed using alkylbenzene derivatives. The pH effect on the electroosmotic flow was also investigated. The open tubular column functionalized with triethanolamine allowed to operate in the anodic electroosmotic flow mode in the system. With anodic electroosmotic flow mode, favorable separations of the amino acids and the nucleosides were successfully achieved with high column efficiens ranging from 142000 to 257000 plates/m. Good repeatability was gained with relative standard deviation of the migration time and peak areas less than 2.2% for run to run (n=5) and less than 3.2% day to day (n=3). Furthermore, real sample applicability of this column to the separation of amino acids in white tea sample was demonstrated.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M6514
3-(Trimethoxysilyl)propyl methacrylate, ≥98%
C10H20O5Si
64208
3-(Trimethoxysilyl)propyl methacrylate, Selectophore
C10H20O5Si
440159
3-(Trimethoxysilyl)propyl methacrylate, 98%
C10H20O5Si
A0230200
Adenosine, European Pharmacopoeia (EP) Reference Standard
C10H13N5O4
PHR1138
Adenosine, Pharmaceutical Secondary Standard; Certified Reference Material
C10H13N5O4
C4654
Cytidine, BioReagent, suitable for cell culture, powder, ≥99%
C9H13N3O5
C122106
Cytidine, 99%
C9H13N3O5
30270
Cytidine, ≥99.0% (HPLC)
C9H13N3O5
G6752
Guanosine, ≥98%
C10H13N5O5
G6264
Guanosine, suitable for cell culture, BioReagent
C10H13N5O5
75903
Guanosine, ≥97.0% (HPLC)
C10H13N5O5
ARK2174
Triethanolamine, PharmaGrade, USP, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production.
C6H15NO3
90278
Triethanolamine, BioUltra, ≥99.5% (GC)
C6H15NO3
90279
Triethanolamine, ≥99.0% (GC)
C6H15NO3
33729
Triethanolamine, puriss. p.a., ≥99% (GC)
C6H15NO3
50926
Triethanolamine, analytical reference material
C6H15NO3
T58300
Triethanolamine, reagent grade, 98%
C6H15NO3
T2545000
Trolamine, European Pharmacopoeia (EP) Reference Standard
C6H15NO3
U3750
Uridine, ≥99%
C9H12N2O6
U3003
Uridine, powder, BioReagent, suitable for cell culture
C9H12N2O6
U6381
Uridine, BioUltra, ≥99%
C9H12N2O6