EMAIL THIS PAGE TO A FRIEND
Molecular cancer therapeutics
Preclinical characterization of RSM-932A, a novel anticancer drug targeting the human choline kinase alpha, an enzyme involved in increased lipid metabolism of cancer cells.
PMID 25487918
Abstract
Choline kinase α (CHKA; here designated as ChoKα) is the first enzyme in the CDP-choline pathway, implicated in phospholipids metabolism. It is overexpressed in several human tumors such as breast, lung, bladder, colorectal, prostate, ovary, and liver. The overexpression of ChoKα has oncogenic potential and synergizes with other known oncogenes. It has been proposed as a novel cancer drug target with a distinct mechanism of action. We have generated a set of ChoKα inhibitors with potent in vitro antiproliferative and in vivo antitumoral activity against human xenografts in mice, showing high efficacy with low toxicity profiles. Among these inhibitors, RSM-932A has been chosen for further clinical development due to its potent antiproliferative activity in vitro against a large variety of tumor-derived cell lines, a potent in vivo anticancer activity, and lack of toxicity at the effective doses. Here, we provide the preclinical evidence to support the use of RSM-932A as a good candidate to be tested in clinical trials as the "first in humans" drug targeting ChoKα.