EMAIL THIS PAGE TO A FRIEND
American journal of ophthalmology
Confocal microscopy analysis of corneal changes after photorefractive keratectomy plus cross-linking for keratoconus: 4-year follow-up.
PMID 25488640
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C9879 Collagen from bovine achilles tendon, powder, suitable for substrate for collagenase
C4387
Collagen from bovine achilles tendon, powder, suitable for substrate for collagenase
C7806 Collagen from bovine nasal septum, Bornstein and Traub Type II, powder
C1188 Collagen from bovine tracheal cartilage, Bornstein and Traub Type II, powder
C8919 Collagen from calf skin, Bornstein and Traub Type I, (0.1% solution in 0.1 M acetic acid), aseptically processed, BioReagent, suitable for cell culture
C9791
Collagen from calf skin, Bornstein and Traub Type I, solid, BioReagent, suitable for cell culture
C3511
Collagen from calf skin, Bornstein and Traub Type I (Sigma Type III), solid
C9301 Collagen from chicken sternal cartilage, Type II (Miller), powder, BioReagent, suitable for cell culture
C0543 Collagen from Engelbreth-Holm-Swarm murine sarcoma basement membrane, Type IV (Miller), lyophilized powder (from 0.2 μm-filtered solution), BioReagent, suitable for cell culture
C7521
Collagen from human placenta, Bornstein and Traub Type IV, powder
C4407 Collagen from human placenta, Bornstein and Traub Type III (Sigma Type X), powder
C5533
Collagen from human placenta, Bornstein and Traub Type IV, powder, BioReagent, suitable for cell culture
C7774 Collagen from human placenta, Bornstein and Traub Type I (Sigma Type VIII), powder
C3657 Collagen from human placenta, Bornstein and Traub Type V (Sigma Type IX), powder
C8374
Collagen from human placenta, Bornstein and Traub Type IV, powder
H4417 Collagen from human placenta, Bornstein and Traub Type IV, solution, suitable for cell culture, ≥95% (SDS-PAGE)
C3929 Collagen from kangaroo tail, Bornstein and Traub Type I, powder
C5608 Collagen from rabbit skin, Bornstein and Traub Type I, powder
C7661 Collagen from rat tail, Bornstein and Traub Type I, powder, BioReagent, suitable for cell culture
27666 Collagen from rat tail, for cell biology, soluble
C5483
Collagen human, Bornstein and Traub Type I, acid soluble, powder, ~95% (SDS-PAGE)
C7624
Collagen human, Bornstein and Traub Type I, recombinant, expressed in Nicotiana tabacum (tobacco)
C7999
Collagen human, Bornstein and Traub Type I, recombinant, expressed in Nicotiana tabacum (tobacco)
C8993 Collagen ovine, aqueous solution
C6745 Collagen Type IV from human cell culture, Bornstein and Traub Type IV, 0.3 mg/mL, sterile-filtered, BioReagent, suitable for cell culture
C3867 Collagen, Type I solution from rat tail, BioReagent, suitable for cell culture, sterile-filtered