EMAIL THIS PAGE TO A FRIEND
Journal of immunology (Baltimore, Md. : 1950)
Identification of Aim2 as a sensor for DNA vaccines.
PMID 25488991
Abstract
Recent human study data have re-established the value of DNA vaccines, especially in priming high-level Ag-specific Ab responses, but also raised questions about the mechanisms responsible for such effects. Whereas previous reports have shown involvement of downstream signaling molecules in the innate immune system, the current study investigated the role of absent in melanoma 2 (Aim2) as a sensor for DNA vaccines. The Aim2 inflammasome directs maturation of the proinflammatory cytokines IL-1β and IL-18 and an inflammatory form of cell death called pyroptosis. Both the humoral and cellular Ag-specific adaptive responses were significantly reduced in Aim2-deficient mice in an IL-1β/IL-18-independent manner after DNA vaccination. Surprisingly, Aim2-deficient mice also exhibited significantly lower levels of IFN-α/β at the site of injection. These results indicate a previously unreported link between DNA vaccine-induced pyroptotic cell death and vaccine immunogenicity that is instrumental in shaping the Ag-specific immune response to DNA vaccines.