EMAIL THIS PAGE TO A FRIEND
Molecular vision
Long-term preservation of cone photoreceptors and visual acuity in rd10 mutant mice exposed to continuous environmental enrichment.
PMID 25489227
Abstract
In human patients and animal models of retinitis pigmentosa (RP), a gradual loss of rod photoreceptors and decline in scotopic vision are the primary manifestations of the disease. Secondary death of cones and gradual, regressive remodeling of the inner retina follow and progress at different speeds according to the underlying genetic defect. In any case, the final outcome is near-blindness without a conclusive cure yet. We recently reported that environmental enrichment (EE), an experimental manipulation based on exposure to enhanced motor, sensory, and social stimulation, when started at birth, exerts clear beneficial effects on a mouse model of RP, by slowing vision loss. The purpose of this study was to investigate in the same mouse the long-term effects of chronic exposure to an EE and assess the outcome of this manipulation on cone survival, inner retinal preservation, and visual behavior. Two groups of rd10 mutant mice were maintained in an EE or standard (ST) laboratory conditions up to 1 year of age. Then, retinal preservation was assessed with immunocytochemistry, confocal microscopy examination, cone counts, and electron microscopy of the photoreceptor layer, while visual acuity was tested behaviorally with a Prusky water maze. rd10 mice are a model of autosomal recessive RP with a typical rod-cone, center to the periphery pattern of photoreceptor degeneration. They carry a mutation of the rod-specific phosphodiesterase gene and undergo rod death that peaks at around P24, while cone electroretinogram (ERG) is extinct by P60. We previously showed that early exposure to an EE efficiently delays photoreceptor degeneration in these mutants, extending the time window of cone viability and cone-mediated vision well beyond the phase of maximum rod death. Here we find that a maintained EE can delay the degeneration of cones even in the long term. Confocal and electron microscopy examination of the retinas of the rd10 EE and ST mice at 1 year of age showed major degeneration of the photoreceptor layer in both experimental groups, with small clusters of photoreceptors persisting in the peripheral retina. These vestigial cells were positive for L and M opsins and cone arrestin and represented the residual population of cones. In the retinas of the EE mice, cones were more numerous and less remodeled than in the ST counterparts, albeit virtually devoid of outer segments, as confirmed with electron microscopy (EM) observations. Cone counting in retinal whole mounts showed that rd10 EE mice at 1 year had almost three times as many surviving cones (34,000±4,000) as the ST control mice (12,700±1,800), t test p=0.003. Accordingly, the rd10 EE mice at 1 year of age were still capable of performing the visual water task in photopic conditions, showing a residual visual acuity of 0.138±0 cycles/degree. This ability was virtually absent in the rd10 ST age-matched mice (0.063±0.014), t test, p=0.029. No major differences were detected in the morphology of the neurons of the inner retina between the two experimental groups. The approaches used to test the effects of an EE were consistent in showing significantly better preservation of cones and measurable visual acuity in 1-year-old rd10 EE mice. We therefore confirm and extend previous findings that showed an EE is an effective, minimally invasive tool for promoting long-lasting retinal protection in experimental models of RP.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AB15282
Anti-Cone Arrestin Antibody, 100 µg, Anti-Cone Arrestin, Cat. No. AB15282, is a highly specific rabbit polyclonal antibody that targets Arrestin-C and has been tested in Immunohistochemistry (Paraffin) and Western Blotting.
AB5404
Anti-Neurturin Antibody, 50 µL, Anti-Neurturin Antibody detects level of Neurturin & has been published & validated for use in IH.
AB5407
Anti-Opsin Antibody, blue, 100 µg, This Anti-Opsin Antibody, blue is validated for use in IH(P) for the detection of Opsin.
A9979
Anti-Aly antibody, Mouse monoclonal, clone 11G5, purified from hybridoma cell culture
SAB2701734
Anti-cdk7 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA007932
Anti-CDK7 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV03009
Anti-CDK7 antibody produced in rabbit, IgG fraction of antiserum
SAB2104125
Anti-CDK7 antibody produced in rabbit, affinity isolated antibody
SAB4500060
Anti-CDK7 antibody produced in rabbit, affinity isolated antibody
SAB1405672
Anti-CTBP2 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2701876
Anti-ctbp2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2701877
Anti-ctbp2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
AV50670
Anti-CTBP2 antibody produced in rabbit, affinity isolated antibody
HPA023559
Anti-CTBP2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1
HPA023564
Anti-CTBP2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2
SAB2105753
Anti-CTBP2 antibody produced in rabbit, affinity isolated antibody
SAB4300489
Anti-KIT (Ab-721) antibody produced in rabbit, affinity isolated antibody
SAB4300593
Anti-KIT (Ab-936) antibody produced in rabbit, affinity isolated antibody
GW21114 Anti-KIT antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB1412042
ANTI-KIT antibody produced in mouse, clone 1G1, purified immunoglobulin, buffered aqueous solution
SAB1412043
ANTI-KIT antibody produced in mouse, clone 5F6, purified immunoglobulin, buffered aqueous solution
SAB1412044
ANTI-KIT antibody produced in mouse, clone 5A11, purified immunoglobulin, buffered aqueous solution
SAB1412045
ANTI-KIT antibody produced in mouse, clone 2C3, purified immunoglobulin, buffered aqueous solution
SAB1412046
ANTI-KIT antibody produced in mouse, clone 1D3, purified immunoglobulin, buffered aqueous solution
SAB1412140
ANTI-KIT antibody produced in mouse, clone 4F7, purified immunoglobulin, buffered aqueous solution
SAB1412141
ANTI-KIT antibody produced in mouse, clone 3A8, purified immunoglobulin, buffered aqueous solution
HPA004471
Anti-KIT antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1411601
Anti-MAP3K14 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1306410
ANTI-NIK (CENTER) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
PRS1129
Anti-NIK antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution