EMAIL THIS PAGE TO A FRIEND
International journal of pharmaceutics
pH-responsive, lysine-based hydrogels for the oral delivery of a wide size range of molecules.
PMID 25490181
Abstract
Hydrogels synthesized from poly(l-lysine isophthalamide) (PLP) crosslinked with l-lysine methyl ester were investigated as drug delivery systems for a wide size range of molecules (0.3-2000kDa). PLP is an anionic, pseudo-peptidic polymer that is an ideal hydrogel backbone due to its pH-responsiveness and amphiphilicity. Drug loading and release were evaluated for various model drugs: hydrophobic fluorescein (Mw=332Da) and hydrophilic fluorescein isothiocyanate-dextran (FITC-Dex Mw=10kDa, 150kDa, 500kDa, and 2000kDa). Weight incorporation was high, up to 22.8±3.1%. Release after 24h in pH 7.4 was in the range from 70.4±1.2% to 91.6±0.8% for all model drugs. In contrast, drug release after 24h in pH 3.0 was significantly lower, less than 8% for fluorescein, 500kDa, and 2000kDa FITC-Dex. Thus, the adaptability of these novel hydrogels to both hydrophobic and hydrophilic molecules, spanning a wide size range, suggests their use as a platform delivery system. This is also the first known hydrogel system for the oral delivery of payloads larger than 70kDa, which, combined with triggered release in response to pH changes along the gastrointestinal tract, indicates that these hydrogels have promising applications in oral drug delivery.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
59210
Isophthaloyl chloride, purum, ≥98.0% (AT)
C8H4Cl2O2
I19403
Isophthaloyl chloride, ≥99%
C8H4Cl2O2
L0645
L-Lysine methyl ester dihydrochloride
C7H16N2O2 · 2HCl
P3657
Pyridoxal 5′-phosphate hydrate, powder, BioReagent, suitable for cell culture
C8H10NO6P · xH2O
W302600
Sodium citrate dihydrate, ≥99%, FG
C6H5Na3O7 · 2H2O
S4641
Sodium citrate tribasic dihydrate, ACS reagent, ≥99.0%
C6H5Na3O7 · 2H2O
C7254
Sodium citrate tribasic dihydrate, ≥98%
C6H5Na3O7 · 2H2O
C8532
Sodium citrate tribasic dihydrate, for molecular biology, ≥99%
C6H5Na3O7 · 2H2O
C3434
Sodium citrate tribasic dihydrate, BioXtra, ≥99.0% (titration)
C6H5Na3O7 · 2H2O
71402
Sodium citrate tribasic dihydrate, BioUltra, for molecular biology, ≥99.5% (NT)
C6H5Na3O7 · 2H2O
C0909
Sodium citrate tribasic dihydrate, suitable for amino acid analysis, ≥98% (titration), powder
C6H5Na3O7 · 2H2O
C5920
Sodium citrate tribasic dihydrate, insect cell culture tested
C6H5Na3O7 · 2H2O
71404
Sodium citrate tribasic dihydrate, BioUltra, ≥99.0% (NT)
C6H5Na3O7 · 2H2O
71406
Sodium citrate tribasic dihydrate, purum p.a., ≥99.0% (NT)
C6H5Na3O7 · 2H2O
71409
Sodium citrate tribasic dihydrate, tested according to Ph.Eur.
C6H5Na3O7 · 2H2O
S1804
Trisodium citrate dihydrate, meets USP testing specifications
C6H5Na3O7 · 2H2O