EMAIL THIS PAGE TO A FRIEND
Journal of molecular biology
Structure of an APC3-APC16 complex: insights into assembly of the anaphase-promoting complex/cyclosome.
PMID 25490258
Abstract
The anaphase-promoting complex/cyclosome (APC/C) is a massive E3 ligase that controls mitosis by catalyzing ubiquitination of key cell cycle regulatory proteins. The APC/C assembly contains two subcomplexes: the "Platform" centers around a cullin-RING-like E3 ligase catalytic core; the "Arc Lamp" is a hub that mediates transient association with regulators and ubiquitination substrates. The Arc Lamp contains the small subunits APC16, CDC26, and APC13, and tetratricopeptide repeat (TPR) proteins (APC7, APC3, APC6, and APC8) that homodimerize and stack with quasi-2-fold symmetry. Within the APC/C complex, APC3 serves as center for regulation. APC3's TPR motifs recruit substrate-binding coactivators, CDC20 and CDH1, via their C-terminal conserved Ile-Arg (IR) tail sequences. Human APC3 also binds APC16 and APC7 and contains a >200-residue loop that is heavily phosphorylated during mitosis, although the basis for APC3 interactions and whether loop phosphorylation is required for ubiquitination are unclear. Here, we map the basis for human APC3 assembly with APC16 and APC7, report crystal structures of APC3Δloop alone and in complex with the C-terminal domain of APC16, and test roles of APC3's loop and IR tail binding surfaces in APC/C-catalyzed ubiquitination. The structures show how one APC16 binds asymmetrically to the symmetric APC3 dimer and, together with biochemistry and prior data, explain how APC16 recruits APC7 to APC3, show how APC3's C-terminal domain is rearranged in the full APC/C assembly, and visualize residues in the IR tail binding cleft important for coactivator-dependent ubiquitination. Overall, the results provide insights into assembly, regulation, and interactions of TPR proteins and the APC/C.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A5508
Adenosine 5′-phosphosulfate sodium salt, ≥85%
C10H14N5O10PS
SAB1400382
Anti-ANAPC10 antibody produced in mouse, IgG fraction of antiserum, buffered aqueous solution
SAB2900318
Anti-ANAPC10 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA044547
Anti-ANAPC10 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1410084
Anti-ANAPC10 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
HPA037815
Anti-ANAPC16 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV41605
Anti-ANAPC7 antibody produced in rabbit, affinity isolated antibody
SAB3500186
Anti-APC10 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB3500460
Anti-APC3 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB3500309
Anti-APC7 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA028129
Anti-CDC27 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4503747
Anti-H-NUC antibody produced in rabbit, affinity isolated antibody
SAB2700220
Anti-SCARA3 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA047386
Anti-SCARA3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2106762
Anti-SCARA3 antibody produced in rabbit, affinity isolated antibody
C7104
Monoclonal Anti-Cdc27 antibody produced in mouse, ~2 mg/mL, clone AF3.1, purified immunoglobulin, buffered aqueous solution
SAB5300351
Monoclonal Anti-CDC27 antibody produced in mouse, clone 5C12, ascites fluid
WH0051435M1
Monoclonal Anti-SCARA3 antibody produced in mouse, clone 3A2, purified immunoglobulin, buffered aqueous solution