EMAIL THIS PAGE TO A FRIEND
Materials science & engineering. C, Materials for biological applications
Influence of Cu content on the cell biocompatibility of Ti-Cu sintered alloys.
PMID 25491971
Abstract
The cell toxicity and the cell function of Ti-Cu sintered alloys with different Cu contents (2, 5, 10 and 25 wt.%, respectively) have been investigated in comparison with commercial pure titanium in order to assess the influence of Cu content on the cell biocompatibility of the Ti-Cu alloys. The cytotoxicity was studied by examining the MG63 cell response by CCK8 assessment. The cell morphology was evaluated by acridine orange/ethidium bromide (AO/EB) fluorescence and observed under scanning electronic microscopy (SEM). The cell function was monitored by measuring the AKP activity. It has been shown by the AO/EB morphology results that the cell death on both cp-Ti sample and Ti-Cu samples is due to apoptosis rather than necrosis. Although more apoptotic cells were found on the Ti-2Cu and Ti-5Cu samples, no evidence of Cu content dependent manner of apoptosis has been found. SEM observation indicated very good cell adhesion and spread on the cp-Ti sample and the Ti-Cu samples with different Cu contents. CCK8 results displayed that increase in the Cu content in Ti-Cu alloys does not bring about any difference in the cell viability. In addition, AKP test results indicated that no difference in the differentiation of MG63 was found between the cp-Ti and the Ti-Cu samples and among the Ti-Cu samples. All results indicated that Ti-Cu alloys exhibit very good cell biocompatibility and the Cu content up to 25 wt.% in the Ti-Cu alloys has no influence on the cell proliferation and differentiation.