EMAIL THIS PAGE TO A FRIEND
Nihon yakurigaku zasshi. Folia pharmacologica Japonica
[Pharmacological profile and clinical findings of a new phosphate binder, ferric citrate hydrate (Riona® Tablets)].
PMID 25492366