EMAIL THIS PAGE TO A FRIEND
Dalton transactions (Cambridge, England : 2003)
Towards the elaboration of new gold-based optical theranostics.
PMID 25492395
Abstract
Four new red BODIPY-gold(I) theranostic compounds were synthesized. Some of them were vectorized by tethering a biovector (glucose or bombesin derivatives) to the metallic center. Their photophysical properties were studied. Additionally, their cytotoxicity was examined on different cancer cell lines and on a normal cell line, they were tracked in vitro by fluorescence detection, and their uptake was evaluated by ICP-MS measurements.