EMAIL THIS PAGE TO A FRIEND
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine
A polysaccharide from mushroom Huaier retards human hepatocellular carcinoma growth, angiogenesis, and metastasis in nude mice.
PMID 25492485
Abstract
Mushroom Huaier has become a focus of interest in the treatment of hepatocellular carcinoma (HCC). Presently, we isolated and purified one polysaccharide from this mushroom. This study aimed to investigate the effects of SP1 on tumor growth and metastasis in a HCC xenograft model and explore its possible mechanism of action. Our results showed that SP1 not only significantly inhibited the proliferation of SMMC-7721 cells in vitro at the concentration ranging from 0 to 800xa0μg/ml but also suppressed the HCC tumor growth and metastatic nodules to the lung in SMMC-7721-bearing mice by oral administration at three doses of 30, 60, and 120xa0mg/kg. Concomitantly, immunohistochemistry analysis of tumor tissues identified that SP1 administration at three doses significantly inhibited the in vivo cancer cell proliferation and microvessel density (MVD) formation, evidenced by a low proliferating cell nuclear antigen (PCNA) and CD34 expression, but increased the percentage of terminal deoxynucleotidyl transferase-mediated dUTP nick end labeling (TUNEL)-positive cells. Keeping in line with this observation, SP1 treatment decreased serum matrix metalloproteinase (MMP) 2 and vascular endothelial growth factor (VEGF) levels, downregulated the protein expression of hypoxia-inducible factor (HIF)-1alpha, VEGF, MMP2, bcl-2, N-cadherin, signal transducer and activator of transcription 3 (STAT3), and metadherin (MTDH), and upregulated bax and NE-cadherin protein expression in tumor tissues. Taken together, our data suggest that SP1 appears to be a promising chemopreventive agent for the tumorigenesis and metastasis in patients with HCC, especially at advanced stages.