EMAIL THIS PAGE TO A FRIEND
Molecular therapy : the journal of the American Society of Gene Therapy
Functional aspects of intrahepatic hepatitis B virus-specific T cells induced by therapeutic DNA vaccination.
PMID 25492563
Abstract
Current therapies for the hepatitis B virus (HBV), a major cause of severe liver disease, suppress viral replication but replication rebounds if therapy is withdrawn. It is widely accepted that immune activation is needed to control replication off-therapy. To specifically activate T cells crossreactive between the hepatitis B core and e antigens (HBcAg/HBeAg) in chronically infected patients, we developed a therapeutic vaccine candidate. The vaccine encompass codon-optimized HBcAg and IL-12 expressing plasmids delivered using targeted high-pressure injection combined with in vivo electroporation. One dose of the vaccine primed a B-cell-independent polyfunctional T-cell response, in wild-type, and in HBeAg-transgenic mice with an impaired ability to respond to HBc/eAg. The response peaked at 2 weeks and contracted at week 6 after vaccination. Coadministration of IL-12 improved antibody levels, and T-cell expansion and functionality. The vaccine primed T cells that, 2 weeks after a single dose, cleared hepatocytes transiently expressing HBcAg in vaccinated wild-type and HBeAg-transgenic mice. However, 4 weeks later, these functional responses were lost. Booster doses after 8-12 weeks effectively restored function and expansion of the rapidly contracting T cells. Thus, this vaccine strategy primes functional HBcAg-specific T cells in a host with dysfunctional response to HBV.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
89458
Aphidicolin, analytical standard
C20H34O4