EMAIL THIS PAGE TO A FRIEND
Nature communications
microTSS: accurate microRNA transcription start site identification reveals a significant number of divergent pri-miRNAs.
PMID 25492647
Abstract
A large fraction of microRNAs (miRNAs) are derived from intergenic non-coding loci and the identification of their promoters remains 'elusive'. Here, we present microTSS, a machine-learning algorithm that provides highly accurate, single-nucleotide resolution predictions for intergenic miRNA transcription start sites (TSSs). MicroTSS integrates high-resolution RNA-sequencing data with active transcription marks derived from chromatin immunoprecipitation and DNase-sequencing to enable the characterization of tissue-specific promoters. MicroTSS is validated with a specifically designed Drosha-null/conditional-null mouse model, generated using the conditional by inversion (COIN) methodology. Analyses of global run-on sequencing data revealed numerous pri-miRNAs in human and mouse either originating from divergent transcription at promoters of active genes or partially overlapping with annotated long non-coding RNAs. MicroTSS is readily applicable to any cell or tissue samples and constitutes the missing part towards integrating the regulation of miRNA transcription into the modelling of tissue-specific regulatory networks.