EMAIL THIS PAGE TO A FRIEND
The EMBO journal
Autophagy mediates nonselective RNA degradation in starving yeast.
PMID 25492883
Abstract
During nitrogen starvation, a nonselective bulk degradation of cytosolic proteins and organelles including ribosomes, termed macro‐autophagy (hereafter termed autophagy), is induced. The precise mechanism of RNA degradation by this pathway has not been yet elucidated. In this issue of the The EMBO Journal, Huang et al characterize an autophagy‐dependent RNA catabolism in yeast and identify the enzymes responsible for the degradation process.