EMAIL THIS PAGE TO A FRIEND
Nucleic acid therapeutics
One-staged aptamer-based isolation and application of endothelial progenitor cells in a porcine myocardial infarction model.
PMID 25494449
Abstract
A multitude of stem cell types has been extensively studied and used for myocardial regenerative therapy. Amongst these endothelial progenitor cells form a promising source. In our present study, we investigated a one-staged approach for isolation and application of autologous endothelial progenitor cells in a pig model of myocardial infarction. Endothelial progenitor cell isolation was performed by immediately preprocedural bone marrow aspiration and consecutive positive selection by aptamer-based magnetic cell sorting. Animals were divided in three groups receiving endothelial progenitor cells, saline, or no intramyocardial injection respectively. Postprocedural follow-up consisted of weekly echocardiographic evaluations. Postmortem histological analysis after four weeks focused on detection of transplanted PKH26-positive endothelial progenitor cells and neovascularization within the infarcted myocardium. A significant difference in left ventricular ejection fraction could not be shown between the three groups. PKH26-stained endothelial progenitor cells could be found in the endothelial progenitor cells transplanted group, although detection was scarce. Large-sized capillaries were found to be significantly more in endothelial progenitor cells treated myocardium. The one-stage approach of endothelial progenitor cells isolation and application presented herein offers a new therapeutic concept. Even though a beneficial impact on myocardial function could not be assessed, increased neovascularization may indicate positive effects on remodeling processes. Being able to harvest endothelial progenitor cells right before application provides a wider scope of action in urgent cases.