EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Endo-selective Pd-catalyzed silyl methyl Heck reaction.
PMID 25494921
Abstract
A palladium (Pd)-catalyzed endo-selective Heck reaction of iodomethylsilyl ethers of phenols and aliphatic alkenols has been developed. Mechanistic studies reveal that this silyl methyl Heck reaction operates via a hybrid Pd-radical process and that the silicon atom is crucial for the observed endo selectivity. The obtained allylic silyloxycycles were further oxidized into (Z)-alkenyldiols.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
555142
1-Diphenylphosphino-1′-(di-tert-butylphosphino)ferrocene
C30H36FeP2