EMAIL THIS PAGE TO A FRIEND
American journal of respiratory and critical care medicine
Bridging the gap: merging clinical and inflammatory phenotypes with epithelial gene expression profiles in asthma.
PMID 25496099