EMAIL THIS PAGE TO A FRIEND
American journal of respiratory and critical care medicine
Using exhaled nitric oxide and serum periostin as a composite marker to identify severe/steroid-insensitive asthma.
PMID 25496105