EMAIL THIS PAGE TO A FRIEND
The Journal of pharmacy and pharmacology
Regioselective glucuronidation of gingerols by human liver microsomes and expressed UDP-glucuronosyltransferase enzymes: reaction kinetics and activity correlation analyses for UGT1A9 and UGT2B7.
PMID 25496264
Abstract
To determine the reaction kinetics for regioselective glucuronidation of gingerols (i.e. 6-, 8- and 10-gingerol) by human liver microsomes and expressed UDP-glucuronosyltransferase (UGT) enzymes, and to identify the main UGT enzymes involved in regioselective glucuronidation of gingerols. The rates of glucuronidation were determined by incubating the gingerols with uridine diphosphoglucuronic acid-supplemented microsomes. Kinetic parameters were derived by fitting an appropriate model to the data. Activity correlation analyses were performed to identify the main UGT enzymes contributing to hepatic metabolism of gingerols. Glucuronidation at the 4'-OH group was much more favoured than that at 5-OH. The degree of position preference was compound-dependent; the catalytic efficiency ratios of 4'-O- to 5-O-glucuronidation were 9.1, 19.7 and 2.9 for 6-, 8- and 10-gingerol, respectively. UGT1A8 (an intestinal enzyme), UGT1A9 and UGT2B7 were the enzymes showing the highest activity towards gingerols. Formation of 5-O-glucuronide was mainly catalysed by UGT1A9. UGT2B7 was the only enzyme that generated glucuronides at both 4'-OH and 5-OH sites, although a strong position preference was observed with 4'-OH (≥80.2%). Further, activity correlation analyses indicated that UGT2B7 and UGT1A9 were primarily responsible for 4'-O-glucuronidation and 5-O-glucuronidation of gingerols in the liver, respectively. Gingerols were metabolized by multiple hepatic and gastrointestinal UGT enzymes. Also, UGT1A9 and 2B7 were the main contributors to regioselective glucuronidation of gingerols in the liver.