EMAIL THIS PAGE TO A FRIEND
Journal of controlled release : official journal of the Controlled Release Society
In vitro dry powder inhaler formulation performance considerations.
PMID 25497311
Abstract
It has long been desired to match airflow conditions during formulation evaluation to those of relevance to lung deposition. In this context several strategies have been adopted involving sampling at different: flow rate (without consideration of flow conditions, e.g. shear, Reynolds number, work function); pressure drop (with and without consideration of flow conditions) and; flow rate and pressure drop. Performance testing has focused on the influence of these sampling conditions on delivered dose uniformity and aerodynamic particle size distribution. However, in order to be physiologically relevant it is also important to know when the drug was delivered with respect to initiation of airflow as variation in this parameter would influence lung deposition. A light obscuration method of detecting the dose delivered from a dry powder inhaler while sampling for aerodynamic particle size distributions (APSD) by inertial impaction has been developed. Four formulations of albuterol sulfate and budesonide in sieved and milled lactose, respectively, were dispersed and their rate of delivery monitored. The differences observed have the potential to impact the site of delivery in the lungs. The rate of delivery of drug is clearly an important companion measurement to delivered dose and APSD if the intent is to predict the similarity of in vivo performance of dry powder inhaler products.