EMAIL THIS PAGE TO A FRIEND
Vaccine
Preparation of pure, high titer, pseudoinfectious Flavivirus particles by hollow fiber tangential flow filtration and anion exchange chromatography.
PMID 25498209
Abstract
Purification of enveloped viruses such as live flavivirus vaccine candidates poses a challenge as one must retain viral infectivity to preserve immunogenicity. Here we describe a laboratory-scale purification procedure for two replication defective (single-cycle) flavivirus variants for use in a pre-clinical setting. The two step purification scheme based on hollow fiber tangential flow filtration (TFF) followed by anion exchange chromatography using convective interaction media (CIM(®)) monoliths results in a ∼60% recovery of infectious virus titer and can be used to prepare nearly homogenous, highly purified vaccine viruses with titers as high as 1×10(9) focus forming units per mL. Flavivirus virions prepared by this method are 2 and 3 orders of magnitude more pure with respect to dsDNA and BHK host cell proteins, respectively, as compared to the raw feed stream.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB3701024 Anti-Mouse IgG (Fc specific)-Alkaline Phosphatase antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB3701038 Anti-Mouse IgG (γ-chain specific)-Alkaline Phosphatase antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB3701117 Anti-Mouse IgG (H+L)-Alkaline Phosphatase antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB3701107 Anti-Mouse IgG (H+L)-Alkaline Phosphatase antibody produced in donkey, affinity isolated antibody, buffered aqueous solution
SAB3701085 Anti-Mouse IgG (H+L)-Alkaline Phosphatase antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB3701096 Anti-Mouse IgG (H+L)-Alkaline Phosphatase antibody produced in sheep, affinity isolated antibody, buffered aqueous solution
A1902 Anti-Mouse IgG (whole molecule)−Alkaline Phosphatase antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
A2418 Anti-Mouse IgG (whole molecule)−Alkaline Phosphatase antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB3701006 Anti-Mouse IgG F(ab′)2, highly cross adsorbed-Alkaline Phosphatase antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB3701005 Anti-Mouse IgG F(ab′)2-Alkaline Phosphatase antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution