EMAIL THIS PAGE TO A FRIEND
Molecular immunology
TIPE2 protein negatively regulates HBV-specific CD8⁺ T lymphocyte functions in humans.
PMID 25499447
Abstract
Cytotoxic T cell-mediated killing of virus-infected hepatocytes is an important pathogenic process of hepatitis B. However, its underlying molecular mechanisms are not fully understood. TNFAIP8L2 (TIPE2) is a newly described anti-inflammatory protein that is essential for maintaining immune homeostasis. In this study, we found that the protein levels of TIPE2 in PBMCs of hepatitis B patients were significantly reduced and negatively correlated with the sera values of aminotransferases. Importantly, TIPE2 protein was downregulated preferentially in cytotoxic CD8(+) T cells, not CD4(+) helper T cells. The CD8(+) T cells with low TIPE2 expression were more activated and produced higher levels of perforin, granzyme B, and IFN-γ. As a result, their cytolytic activity was markedly enhanced. Interestingly, HBc18-27 peptide stimulation could reduce TIPE2 expression in PBMCs. These results indicate that TIPE2 plays an important role in regulating HBV-specific CD8(+) T cell functions in patients with hepatitis B.