EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
A BODIPY-luminol chemiluminescent resonance energy-transfer (CRET) cassette for imaging of cellular superoxide.
PMID 25500943
Abstract
Spectroscopic and in cellulo studies are here reported on the very first BODIPY-luminol chemiluminescent resonance energy-transfer (CRET) cassette where the luminol CL agent is covalently linked to the BODIPY energy-transfer acceptor in a molecular dyad. The efficiency of intramolecular CRET investigated for the BODIPY-luminol dyad was found to be 64% resulting in a dual emissive response. Successful in cellulo biochemiluminescence via CRET was achieved in PMA activated splenocytes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D60400
2,3-Dichloro-5,6-dicyano-p-benzoquinone, 98%
C8Cl2N2O2
266620
Boron, crystalline, 1 cm, 99.7% trace metals basis
B
333360
Boron, crystalline, −60 mesh, 99% trace metals basis
B
332445
Boron, amorphous, powder, (submicron)
B
15580
Boron, ≥95% (boron), amorphous powder
B
GF84487674
Boron, microfoil, disks, 10mm, thinness 0.1μm, specific density 23.5μg/cm2, permanent mylar 3.5μm support, 99.6%
B
GF19976080
Boron, microfoil, disks, 25mm, thinness 0.1μm, specific density 23.5μg/cm2, permanent mylar 3.5μm support, 99.6%
B
GF81914834
Boron, monofilament, 100m, diameter 0.1mm
B
GF54382888
Boron, monofilament, 10m, diameter 0.1mm
B
GF74687080
Boron, monofilament, 10m, diameter 0.2mm
B
GF40338132
Boron, monofilament, 5m, diameter 0.2mm
B
GF97861247
Boron, monofilament, 20m, diameter 0.1mm
B
GF04144586
Boron, monofilament, 20m, diameter 0.2mm
B
GF32655315
Boron, monofilament, 50m, diameter 0.1mm
B
GF70375833
Boron, monofilament, 50m, diameter 0.2mm
B
GF98835069
Boron, monofilament, 5m, diameter 0.1mm
B
GF39354048
Boron, monofilament, 200m, diameter 0.1mm
B
27815
p-Toluenesulfonic acid monohydrate, ≥99% (calc. to H2O free subst.)
C7H8O3S · H2O
402885
p-Toluenesulfonic acid monohydrate, ACS reagent, ≥98.5%
C7H8O3S · H2O
T35920
p-Toluenesulfonic acid monohydrate, ReagentPlus®, ≥98%
C7H8O3S · H2O
161993
p-Toluenesulfonic acid monohydrate, reagent grade, 98%
C7H8O3S · H2O
89760
p-Toluenesulfonic acid monohydrate, puriss., for acetyl determination, ≥98.5% (T)
C7H8O3S · H2O
89762
p-Toluenesulfonic acid monohydrate, ≥98.0% (T), ≥98.0% (HPLC)
C7H8O3S · H2O