EMAIL THIS PAGE TO A FRIEND
Medical oncology (Northwood, London, England)
Protective effect of ulinastatin in patients with non-small cell lung cancer after radiation therapy: a randomized, placebo-controlled study.
PMID 25502081
Abstract
Radiation-induced lung injury (RILI) is a frequent, sometimes life-threatening complication of radiation therapy for the treatment of lung cancer. The anti-inflammatory role of ulinastatin has been well documented, and the potential application of ulinastatin in management of acute lung injury has been suggested in multiple animal studies. In this article, we described a double-blind, randomized, placebo-controlled study in patients with non-small cell lung cancer. A total of 120 patients were randomized into two groups: the trial group was treated with ulinastatin for 3xa0days prior to and for the first 7xa0days of radiation therapy and the control group was treated with placebo for 10xa0days following the same schedule. The results from follow-up studies showed that the incidence and grade of RILI were significantly lower in the trial group than in the control group. Reduction in pulmonary function from baseline was significantly smaller in the trial group than that in the control group. Production of serum TGF-β1, TNF-α and IL-6 decreased significantly in the trial group promptly following radiation therapy. However, no difference in survival or tumour response rate was found between the two groups. The results indicated that ulinastatin exerted a protective effect on radiation-induced lung injury. Treatment with ulinastatin could be an effective management strategy and greatly improve the clinical efficacy of radiation therapy for patients with lung cancer.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
295116
Carbon monoxide, ≥99.0%
CO
418072
Carbon-12C monoxide, 99.95 atom % 12C
12CO