EMAIL THIS PAGE TO A FRIEND
Neurology
Absence of neurocognitive effect of hepatitis C infection in HIV-coinfected people.
PMID 25503616
Abstract
To investigate the effect of hepatitis C virus (HCV) on neurocognitive performance in chronically HIV-infected patients enrolled in the CNS HIV Antiretroviral Therapy Effects Research (CHARTER) study. A total of 1,582 participants in CHARTER who were tested for HCV antibody underwent neurocognitive testing; serum HCV RNA was available for 346 seropositive patients. Neurocognitive performance was compared in 408 HCV-seropositive and 1,174 HCV-seronegative participants and in a subset of 160 seropositive and 707 seronegative participants without serious comorbid neurologic conditions that might impair neurocognitive performance, using linear regression and taking into account HIV-associated and demographic factors (including IV drug use) and liver function. Neurocognitive performance characterized by global deficit scores and the proportion of individuals who were impaired were the same in the HCV-seropositive and HCV-seronegative groups. In univariable analyses in the entire sample, only verbal domain scores showed small statistically different superior performance in the HCV+ group that was not evident in multivariable analysis. In the subgroup without significant comorbidities, scores in all 7 domains of neurocognitive functioning did not differ by HCV serostatus. Among the HCV-seropositive participants, there was no association between neurocognitive performance and serum HCV RNA concentration. In HIV-infected patients, HCV coinfection does not contribute to neurocognitive impairment, at least in the absence of substantial HCV-associated liver damage, which was not evident in our cohort.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A0325000
Alanine, European Pharmacopoeia (EP) Reference Standard
C3H7NO2