EMAIL THIS PAGE TO A FRIEND
American journal of physiology. Renal physiology
Pharmacological inhibition of galectin-3 protects against hypertensive nephropathy.
PMID 25503732
Abstract
Galectin-3 activation is involved in the pathogenesis of renal damage and fibrogenesis. Limited data are available to suggest that galectin-3-targeted intervention is a potential therapeutic candidate for the prevention of chronic kidney disease. Homozygous TGR(mREN)27 (REN2) rats develop severe high blood pressure (BP) and hypertensive end-organ damage, including nephropathy and heart failure. Male REN2 rats were treated with N-acetyllactosamine [galectin-3 inhibitor (Gal3i)] for 6 wk; untreated REN2 and Sprague-Dawley rats served as controls. We measured cardiac function with echocardiogram and invasive hemodynamics before termination. BP and proteinuria were measured at baseline and at 3 and 6 wk. Plasma creatinine was determined at 6 wk. Renal damage was assessed for focal glomerular sclerosis, glomerular desmin expression, glomerular and interstitial macrophages, kidney injury molecule-1 expression, and α-smooth muscle actin expression. Inflammatory cytokines and extracellular matrix proteinases were quantified by quantitative real-time PCR. Systolic BP was higher in control REN2 rats, with no effect of Gal3i treatment. Plasma creatinine and proteinuria were significantly increased in control REN2 rats; Gal3i treatment reduced both. Renal damage (focal glomerular sclerosis, desmin, interstitial macrophages, kidney injury molecule-1, α-smooth muscle actin, collagen type I, and collagen type III) was also improved by Gal3i. All inflammatory markers (CD68, IL-68, galectin-3, and monocyte chemoattractant protein-1) were elevated in control REN2 rats and attenuated by Gal3i. Markers of extracellular matrix turnover were marginally altered in untreated REN2 rats compared with Sprague-Dawley rats. In conclusion, galectin-3 inhibition attenuated hypertensive nephropathy, as indicated by reduced proteinuria, improved renal function, and decreased renal damage. Drugs binding to galectin-3 may be therapeutic candidates for the prevention of chronic kidney disease.