EMAIL THIS PAGE TO A FRIEND
The Journal of comparative neurology
Feedback in the brainstem: an excitatory disynaptic pathway for control of whisking.
PMID 25503925
Abstract
Sensorimotor processing relies on hierarchical neuronal circuits to mediate sensory-driven behaviors. In the mouse vibrissa system, trigeminal brainstem circuits are thought to mediate the first stage of vibrissa scanning control via sensory feedback that provides reflexive protraction in response to stimulation. However, these circuits are not well defined. Here we describe a complete disynaptic sensory receptor-to-muscle circuit for positive feedback in vibrissa movement. We identified a novel region of trigeminal brainstem, spinal trigeminal nucleus pars muralis, which contains a class of vGluT2+ excitatory projection neurons involved in vibrissa motor control. Complementary single- and dual-labeling with traditional and virus tracers demonstrate that these neurons both receive primary inputs from vibrissa sensory afferent fibers and send monosynaptic connections to facial nucleus motoneurons that directly innervate vibrissa musculature. These anatomical results suggest a general role of disynaptic architecture in fast positive feedback for motor output that drives active sensation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D5637
3,3′-Diaminobenzidine tetrahydrochloride hydrate, ≥96%
C12H14N4 · 4HCl · xH2O
B3503
Benzidine, ≥98.0% (N)
C12H12N2
B1883
Benzidine, ISOPAC®, ≥98.0% (N)
C12H12N2
31614
Benzidine, analytical standard
C12H12N2
12115
Benzidine, purum p.a., ≥98.0% (N)
C12H12N2
N11779
Cacodylic acid, analytical standard
C2H7AsO2
C0125
Cacodylic acid, ≥98%
C2H7AsO2
20835
Cacodylic acid, ≥99.0%
C2H7AsO2
05-0015
Cacodylic acid, SAJ first grade
C2H7AsO2
C3674
Citric acid trisodium salt, anhydrous, ≥98% (GC)
C6H5Na3O7
C5042
Cresyl Violet acetate, certified by the Biological Stain Commission
C18H15N3O3
D9026
Digoxigenin, analytical standard
C23H34O5
Y0000640
Digoxigenin, European Pharmacopoeia (EP) Reference Standard
C23H34O5
613339
Formamide solution, NMR reference standard, 90% in DMSO-d6 (99.9 atom % D), NMR tube size 3 mm × 8 in.
CH3NO
551384
Formamide solution, NMR reference standard, 90% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
CH3NO
612707
Formamide solution, NMR reference standard, 45% in DMSO-d6 (99.9 atom % D), NMR tube size 3 mm × 8 in.
CH3NO
611891
Formamide solution, NMR reference standard, 45% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
CH3NO
61739
N-Lauroylsarcosine, purum p.a., ≥98.0% (GC)
C15H29NO3
L5000
N-Lauroylsarcosine, neat, ≥95%
C15H29NO3
P38135 Phosphate buffered saline, powder, pH 7.4, for preparing 5 L solutions
247642
Xylenes, ACS reagent, ≥98.5% xylenes + ethylbenzene basis
C8H10
320579
Xylenes, ACS reagent, ≥98.5% xylenes + ethylbenzene basis
C8H10
676748
Xylenes, ACS reagent, ≥98.5% xylenes + ethylbenzene basis (ethylbenzene <=25%)
C8H10
534056
Xylenes, histological grade
C8H10
214736
Xylenes, reagent grade
C8H10
33817
Xylenes, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur.
C8H10