EMAIL THIS PAGE TO A FRIEND
General physiology and biophysics
The effect of chronic peripheral nesfatin-1 application on blood pressure in normal and chronic restraint stressed rats: related with circulating level of blood pressure regulators.
PMID 25504061
Abstract
Nesfatin is a peptide secreted by peripheral tissues, central and peripheral nervous system. It is involved in the regulation of homeostasis. Although the effects of nesfatin-1 on nutrition have been studied widely in the literature, the mechanisms of nesfatin-1 action and also relations with other physiological parameters are still not clarified well. We aimed to investigate the effect of peripheral chronic nesfatin-1 application on blood pressure regulation in normal and in rats exposed to restraint immobilization stress. In our study, three month-old male Wistar rats were used. Rats were divided into 4 groups as Control, Stress, Control+Nesfatin-1, Nesfatin-1+Stress. Angiotensinogen, angiotensin converting enzyme 2, angiotensin II, endothelin-1, endothelial nitric oxide synthase, aldosterone, cortisol, nesfatin-1 levels were determined in plasma samples by ELISA. Our results have shown that chronic peripheral nesfatin-1 administration increases blood pressure in normal and in rats exposed to chronic restraint stress. Effect of nesfatin-1 on circulating level of angiotensinogen, angiotensin converting enzyme 2, angiotensin II, endothelin-1, endothelial nitric oxide synthase, aldosterone and cortisol has been identified. We can conclude that elevated high blood pressure after chronic peripheral nesfatin-1 administration in rats exposed to chronic restraint stress may be related to decreased plasma level of endothelial nitric oxide synthase concentration.