EMAIL THIS PAGE TO A FRIEND
Stem cells (Dayton, Ohio)
Snai1 promotes ESC exit from the pluripotency by direct repression of self-renewal genes.
PMID 25504116
Abstract
Although much is known about the pluripotency self-renewal circuitry, the molecular events that lead embryonic stem cells (ESCs) exit from pluripotency and begin differentiation are largely unknown. We found that the zinc finger transcription factor Snai1, involved in gastrulation and epithelial-mesenchymal transition, is already expressed in the inner cell mass of the preimplantation blastocysts. In ESCs, Snai1 does not respond to TGFβ or BMP4 signaling but it is induced by retinoic acid treatment, which induces the binding, on the Snai1 promoter, of the retinoid receptors RARγ and RXRα, the dissociation of the Polycomb repressor complex 2 which results in the decrease of H3K27me3, and the increase of histone H3K4me3. Snai1 mediates the repression of pluripotency genes by binding directly to the promoters of Nanog, Nr5a2, Tcl1, c-Kit, and Tcfcp2l1. The transient activation of Snai1 in embryoid bodies induces the expression of the markers of all three germ layers. These results suggest that Snai1 is a key factor that triggers ESCs exit from the pluripotency state and initiate their differentiation processes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H7904
(Z)-4-Hydroxytamoxifen, ≥98% Z isomer
C26H29NO2
R4643
9-cis-Retinoic acid, ≥98% (HPLC)
C20H28O2