EMAIL THIS PAGE TO A FRIEND
Cardiovascular research
miR-21-3p regulates cardiac hypertrophic response by targeting histone deacetylase-8.
PMID 25504627
Abstract
Growing evidences indicate that microRNAs (miRNAs) are involved in cardiac hypertrophy development. Multiple miRNAs have been identified as diagnostic and prognostic biomarkers of cardiac hypertrophy, as well as potential therapeutic tools. The present study aimed to investigate the functions and regulatory mechanisms of miR-21-3p in cardiac hypertrophy. Decreased expression of miR-21-3p was observed in cardiac hypertrophy induced by transverse aortic constriction (TAC) and angiotensin (Ang) II infusion in mice. To further explore the role of miR-21-3p in cardiac hypertrophy, rAAV-miR-21-3p was administered intravenously in mice. Overexpression of miR-21-3p markedly suppressed TAC-induced cardiac hypertrophy and also blocked Ang II-induced cardiac hypertrophy as determined by cardiac function measurement and biomarker detection. Furthermore, western blot assays showed that histone deacetylase-8 (HDAC8) was silenced by miR-21-3p, and luciferase reporter assays showed that miR-21-3p binds to the 3' UTR of HDAC8. Moreover, re-expression of HDAC8 attenuated miR-21-3p-mediated suppression of cardiac hypertrophy by enhancing phospho-Akt and phospho-Gsk3β expression. Our data reveal a role of miR-21-3p in regulating HDAC8 expression and Akt/Gsk3β pathway, and suggest that modulation of miR-21-3p levels may provide a therapeutic approach for cardiac hypertrophy.