EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
MDA-9/syntenin is essential for factor VIIa-induced signaling, migration, and metastasis in melanoma cells.
PMID 25505176
Abstract
Melanoma differentiation associated gene-9 (MDA-9), also known as syntenin, is a novel gene that positively regulates cancer cell motility, invasion, and metastasis through distinct biochemical and signaling pathways, but how MDA-9/syntenin is regulated in response to signals with the extracellular environment and promotes tumor progression is unclear. We now demonstrate that MDA-9/syntenin is dramatically up-regulated by a combination of rFVIIa and factor F(X) in malignant melanoma. Induction of MDA-9/syntenin in melanoma was found to occur in a thrombin-independent signaling pathway and involves the PAR-1/c-Src/Rho GTPases Rac1 and Cdc42/c-Jun N-terminal kinase axis resulting in the activation of paxillin, NF-κB, and matrix metalloproteinase-2 (MMP-2). MDA-9/syntenin physically interacts with c-Src through its PDZ binding motif following stimulation of melanoma cells with rFVIIa and FX. We also document that induction of this signaling pathway is required for TF·FVIIa·Xa-induced cell migration, invasion, and metastasis by melanoma cells. The present finding uncovers a novel role of MDA-9/syntenin as an important TF·FVIIa·Xa/PAR-1-regulated gene that initiates a signaling circuit essential for cell motility and invasion of metastatic melanoma. In these contexts, targeting TF·FVIIa·Xa and its relevant downstream targets such as MDA-9/syntenin, may represent a novel therapeutic strategy to control the evolution of neoplastic cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
31640
4,4′-Diaminodiphenylmethane, analytical standard
C13H14N2