EMAIL THIS PAGE TO A FRIEND
Human molecular genetics
HSP47 and FKBP65 cooperate in the synthesis of type I procollagen.
PMID 25510505
Abstract
Osteogenesis imperfecta (OI) is a genetic disorder that results in low bone mineral density and brittle bones. Most cases result from dominant mutations in the type I procollagen genes, but mutations in a growing number of genes have been identified that produce autosomal recessive forms of the disease. Among these include mutations in the genes SERPINH1 and FKBP10, which encode the type I procollagen chaperones HSP47 and FKBP65, respectively, and predominantly produce a moderately severe form of OI. Little is known about the biochemical consequences of the mutations and how they produce OI. We have identified a new OI mutation in SERPINH1 that results in destabilization and mislocalization of HSP47 and secondarily has similar effects on FKBP65. We found evidence that HSP47 and FKBP65 act cooperatively during posttranslational maturation of type I procollagen and that FKBP65 and HSP47 but fail to properly interact in mutant HSP47 cells. These results thus reveal a common cellular pathway in cases of OI caused by HSP47 and FKBP65 deficiency.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV36578
Anti-ANXA3 (AB1) antibody produced in rabbit, IgG fraction of antiserum
AV36579
Anti-ANXA3 (AB2) antibody produced in rabbit, IgG fraction of antiserum
HPA013398
Anti-ANXA3 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA013431
Anti-ANXA3 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
E1031
Anti-ERGIC-53/p58 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA051171
Anti-FKBP10 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1305638 ANTI-LC3 (APG8A) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB1306269
ANTI-LC3 (APG8A/B) antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
L8793
Anti-LC3A antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
HPA002320
Anti-LMAN1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV46614
Anti-LMAN1 antibody produced in rabbit, IgG fraction of antiserum
SAB2101351
Anti-LMAN1 antibody produced in rabbit, affinity isolated antibody
HPA007649
Anti-MAP1LC3A antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2101425
Anti-MAP1LC3A antibody produced in rabbit, affinity isolated antibody
SAB4300571
Anti-MAP1LC3A antibody produced in rabbit, affinity isolated antibody
SAB1411514
Anti-MAP1LC3A antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB3501064
Anti-MAP1LC3A antibody produced in rabbit, affinity isolated antibody
SAB1304456
ANTI-PDIA2 (CENTER) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
HPA051692
Anti-PDIA2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA053492
Anti-PDIA2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1405897
Anti-PDIA3 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA002645
Anti-PDIA3 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA003230
Anti-PDIA3 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2107799
Anti-PDIA3 antibody produced in rabbit, affinity isolated antibody
SAB1410582
Anti-PDIA3 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
S5950
Anti-SERPINH1 (404-418) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB1405565
Anti-SERPINH1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA029198
Anti-SERPINH1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
171824
DL-Proline, ReagentPlus®, 99%
C5H9NO2
SAB4200585
Monoclonal Anti-ERGIC-53 antibody produced in mouse, ~1.0 mg/mL, clone ERGIC-3, purified immunoglobulin