EMAIL THIS PAGE TO A FRIEND
The Journal of infectious diseases
Association of Cytokines With Exosomes in the Plasma of HIV-1-Seropositive Individuals.
PMID 25512626
Abstract
Human immunodeficiency virus (HIV)-infected and viremic individuals exhibit elevated levels of plasma cytokines. Here we show that most cytokines are not in free form but appear associated with exosomes that are distinct from virions. Purified exosomes were analyzed to determine the levels of 21 cytokines and chemokines and compared with exosome-depleted plasma. Most cytokines were markedly enriched in exosomes from HIV-positive individuals relative to negative controls and to plasma. Moreover, exposure of naive peripheral blood mononuclear cells to exosomes purified from HIV-positive patients induced CD38 expression on naive and central memory CD4(+) and CD8(+) T cells, probably contributing to inflammation and viral propagation via bystander cell activation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV51471
Anti-AC antibody produced in rabbit, affinity isolated antibody
HPA005468
Anti-ASAH1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB3500293
Anti-ASAH1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2104966
Anti-ASAH1 antibody produced in rabbit, affinity isolated antibody
SAB2104624
Anti-ASAH1, (N-terminal) antibody produced in rabbit, affinity isolated antibody
HPA022132
Anti-CD38 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA010088
Anti-CD63 antibody produced in rabbit, affinity isolated antibody, Prestige Antibodies® Powered by Atlas Antibodies, buffered aqueous glycerol solution
C9993 Anti-CD9 antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB4503606
Anti-CD9 antibody produced in rabbit, affinity isolated antibody
SAB2501357
Anti-DCTN3 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1400426
Anti-DCTN3 antibody produced in mouse, IgG fraction of antiserum, buffered aqueous solution
HPA044905
Anti-DCTN3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2107711
Anti-DCTN3 antibody produced in rabbit, affinity isolated antibody
SAB1400735
Anti-NICN1 antibody produced in mouse, IgG fraction of antiserum, buffered aqueous solution
HPA037550
Anti-NICN1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1406423
Anti-SELL antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
WH0000427M1
Monoclonal Anti-ASAH1 antibody produced in mouse, clone 2C9, purified immunoglobulin, buffered aqueous solution
SAB1403576
Monoclonal Anti-ASAH1 antibody produced in mouse, clone 1A7, purified immunoglobulin, buffered aqueous solution
WH0000952M2
Monoclonal Anti-CD38 antibody produced in mouse, clone 4G3, purified immunoglobulin, buffered aqueous solution
SAB4700170 Monoclonal Anti-CD38 antibody produced in mouse, clone HIT2, purified immunoglobulin, buffered aqueous solution
SAB4700489 Monoclonal Anti-CD62L antibody produced in mouse, clone LT-TD180, purified immunoglobulin, buffered aqueous solution
SAB4700493 Monoclonal Anti-CD62L antibody produced in mouse, clone DREG56, purified immunoglobulin, buffered aqueous solution
SAB4700619 Monoclonal Anti-CD62L antibody produced in mouse, clone IVA94, purified immunoglobulin, buffered aqueous solution
SAB4700590 Monoclonal Anti-CD62L antibody produced in rat, clone MEL-14, purified immunoglobulin, buffered aqueous solution
SAB4700215
Monoclonal Anti-CD63 antibody produced in mouse, clone MEM-259, purified immunoglobulin, buffered aqueous solution
SAB4700092 Monoclonal Anti-CD9 antibody produced in mouse, clone MEM-61, purified immunoglobulin, buffered aqueous solution
SAB4700618 Monoclonal Anti-CD9 antibody produced in mouse, clone IVA50, purified immunoglobulin, buffered aqueous solution
SAB1402143
Monoclonal Anti-CD9 antibody produced in mouse, clone 4A2, purified immunoglobulin, buffered aqueous solution
SAB4700567
Monoclonal Anti-Cd9 antibody produced in rat, clone EM-04, purified immunoglobulin, buffered aqueous solution