EMAIL THIS PAGE TO A FRIEND
The American surgeon
Successful preservation of the mesenteric and bowel circulation with treatment for a ruptured superior mesenteric artery aneurysm using the HyperEye Medical System.
PMID 25513910
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
21980
Cardiogreen, for microscopy
C43H47N2NaO6S2