EMAIL THIS PAGE TO A FRIEND
Environmental monitoring and assessment
The contribution of trees outside forests to national tree biomass and carbon stocks--a comparative study across three continents.
PMID 25514855
Abstract
In contrast to forest trees, trees outside forests (TOF) often are not included in the national monitoring of tree resources. Consequently, data about this particular resource is rare, and available information is typically fragmented across the different institutions and stakeholders that deal with one or more of the various TOF types. Thus, even if information is available, it is difficult to aggregate data into overall national statistics. However, the National Forest Monitoring and Assessment (NFMA) programme of FAO offers a unique possibility to study TOF resources because TOF are integrated by default into the NFMA inventory design. We have analysed NFMA data from 11 countries across three continents. For six countries, we found that more than 10% of the national above-ground tree biomass was actually accumulated outside forests. The highest value (73%) was observed for Bangladesh (total forest cover 8.1%, average biomass per hectare in forest 33.4 t ha(-1)) and the lowest (3%) was observed for Zambia (total forest cover 63.9%, average biomass per hectare in forest 32 t ha(-1)). Average TOF biomass stocks were estimated to be smaller than 10 t ha(-1). However, given the large extent of non-forest areas, these stocks sum up to considerable quantities in many countries. There are good reasons to overcome sectoral boundaries and to extend national forest monitoring programmes on a more systematic basis that includes TOF. Such an approach, for example, would generate a more complete picture of the national tree biomass. In the context of climate change mitigation and adaptation, international climate mitigation programmes (e.g. Clean Development Mechanism and Reduced Emission from Deforestation and Degradation) focus on forest trees without considering the impact of TOF, a consideration this study finds crucial if accurate measurements of national tree biomass and carbon pools are required.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C2764
Activated charcoal, untreated, granular, ≤5 mm
C
C2889
Activated charcoal, untreated, granular, 8-20 mesh
C
161551
Activated charcoal, powder, -100 particle size (mesh), decolorizing
C
292591
Activated charcoal, 4-14 mesh, granular, Norit® PK 3-5
C
242233
Activated charcoal, DARCO®, 4-12 mesh particle size, granular
C
242241
Activated charcoal, DARCO®, 12-20 mesh, granular
C
242268
Activated charcoal, DARCO®, 20-40 mesh particle size, granular
C
242276
Activated charcoal, DARCO®, −100 mesh particle size, powder
C
329428
Activated charcoal, NORIT® ROW 0.8 SUPRA, pellets
C
C3014
Activated charcoal, untreated, granular, 20-60 mesh
C
C3345
Activated charcoal, untreated powder, 100-400 mesh
C
C4386
Activated charcoal, acid-washed with hydrochloric acid
C
C5510
Activated charcoal, acid-washed with phosphoric and sulfuric acids
C
390127
Activated charcoal, purified by resistive heating of graphite
C
05112
Activated charcoal, for GC
C
31616
Activated charcoal, puriss. p.a., powder
C
675326
Activated charcoal, DARCO® KB-G
C
675342
Activated charcoal, DARCO® KB-WJ
C
80485
Activated charcoal, for the determination of AOX, 50-150 μm particle size
C
C7606
Activated charcoal, meets USP testing specifications
C
97876
Activated Charcoal Norit®, Norit CA1, from wood, chemically activated, powder
C
37771
Activated Charcoal Norit®, Norit GAC 1240W, from coal, for potable water processing, steam activated, granular
C
22874
Activated Charcoal Norit®, Norit PK 1-3, from peat, steam activated, granular
C
29204
Activated Charcoal Norit®, Norit RB3, for gas purification, steam activated, rod
C
51127
Activated Charcoal Norit®, Norit RB4C, steam activated, rod, for the purification of gas
C
53663
Activated Charcoal Norit®, Norit SX ultra, from peat, corresponds U.S. Food chemicals codex (3rd Ed.), steam activated and acid washed, highly purified, powder
C
93067
Activated Charcoal Norit®, Norit® SX2, powder, from peat, multi-purpose activated charcoal, steam activated and acid washed
C
484164 Carbon, glassy, spherical powder, 2-12 μm, 99.95% trace metals basis
C
633100 Carbon, nanopowder, <100 nm particle size (TEM)
C
GF29093058
Carbon, chopped fiber, 100g, nominal diameter 0.007mm, fiber length 6mm, grade 34-700
C