EMAIL THIS PAGE TO A FRIEND
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine
Epidermal growth factor receptor variant III renders glioma cancer cells less differentiated by JAGGED1.
PMID 25514871
Abstract
Glioblastoma is a highly aggressive primary brain tumor in which the majority of cancer cells are undifferentiated. One of the most common oncogenic drivers for this malignancy is the epidermal growth factor receptor variant III (EGFRvIII), which lacks a portion of the extracellular ligand-binding domain due to deletion of exons 2-7 of the EGFR gene. EGFRvIII plays a critical role in tumor progression, promoting acquisition of stem cell-like features including an undifferentiated state and therapy resistance. However, the molecular mechanisms by which EGFRvIII contributes to cancer cell aggressiveness remain poorly understood. Here, we show that EGFR expression correlates with JAGGED1 expression in glioblastoma patients. Overexpression of EGFRvIII in glioma cell lines augmented JAGGED1 expression at the transcriptional level through the mitogen-activated protein kinase signaling pathway. Consequently, EGFRvIII overexpression drove partial dedifferentiation of glioma cells, as determined by tumorsphere-forming ability and expression of stem cell markers, through JAGGED1 induction. EGFRvIII-mediated radioresistance, but not chemoresistance, was also modulated by JAGGED1. Taken together, our results provide new insight into the mechanism underlying EGFRvIII-driven glioblastoma aggressiveness.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C0400
Carmustine, ≥98%
C5H9Cl2N3O2