EMAIL THIS PAGE TO A FRIEND
International endodontic journal
Antimicrobial activity of Chlorhexidine, Peracetic acid and Sodium hypochlorite/etidronate irrigant solutions against Enterococcus faecalis biofilms.
PMID 25515403
Abstract
To evaluate the antimicrobial effect of 2.5% sodium hypochlorite alone (NaOCl) and associated with 9% HEBP (NaOCl/HEBP), 2% peracetic acid (PAA) and 2% chlorhexidine (CHX), on the viability of Enterococcus faecalis biofilms attached to dentine. Biofilms of E.xa0faecalis were grown on the surface of dentine blocks for 5xa0days and then exposed to the irrigating solutions for 3xa0min. Distilled water was used as the control. The total biovolume and the percentage of dead cells of the infected dentine were measured by means of confocal microscopy and the live/dead technique. Nonparametric tests were used to determine statistical differences (Pxa0