EMAIL THIS PAGE TO A FRIEND
Magyar onkologia
[PET radiopharmaceuticals: novelties and new possibilities].
PMID 25517442
Abstract
18F-fluoro-deoxyglucose (FDG) can be considered as the "work-horse" of PET/CT and PET/MR imaging modalities. FDG provides insight in the pathophysiology of tumors and metastases from the point of view of sugar consumption. On the other hand, amino acid metabolism, expression of various receptors in the cells or on the surface of the cells, angiogenesis, appearance of hypoxic cells/tissues and apoptosis also participate in the pathophysiological processes and may have importance in determining the treatment strategy for patients or in monitoring the chosen therapy. Many molecules involved can be labeled by (18)F radionuclide but certain metabolisms require (11)C-labelled agents. Molecular imaging is of key importance in cancer research and various metal complexes containing (44)Sc, (64)Cu, (68)Ga, (86)Y, (89)Zr positron emitters can be very useful in this activity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
261246
Scandium, dendritic pieces, purified by distillation, 99.9% trace rare earth metals basis
Sc
261262
Scandium, powder, 99.9% trace rare earth metals basis
Sc
GF21428153
Scandium, foil, not light tested, 25x25mm, thickness 0.0125mm, as rolled, 99%
Sc
GF23527953
Scandium, foil, 24x25mm, thickness 0.12mm, as rolled, 99%
Sc
GF31290207
Scandium, foil, not light tested, 25x25mm, thickness 0.025mm, as rolled, 99%
Sc
GF55591162
Scandium, foil, 50x50mm, thickness 0.1mm, as rolled, 99%
Sc
GF79587994
Scandium, foil, 25x25mm, thickness 0.1mm, as rolled, 99%
Sc
419141
Zirconium, foil, thickness 0.1 mm, 99.98% trace metals basis
Zr
403296
Zirconium, powder, −100 mesh
Zr
267724
Zirconium, rod, diam. 6.35 mm, ≥99% trace metals basis
Zr
267651
Zirconium, sponge, ≥99% trace metals basis
Zr
14602
Zirconium, ≥97.0% Zr + Hf basis, powder
Zr
756385
Zirconium, powder, in water, -325 mesh, 99.5% trace metals basis (excluding Hf)
Zr
GF69451487
Zirconium, foil, 300x300mm, thickness 0.25mm, annealed, 99.2%
Zr
GF27504402
Zirconium, foil, 300x300mm, thickness 0.5mm, annealed, 99.2%
Zr
GF74069837
Zirconium, foil, 50x50mm, thickness 0.125mm, annealed, 99.2%
Zr
GF13629483
Zirconium, foil, 50x50mm, thickness 0.1mm, annealed, 99.2%
Zr
GF87053474
Zirconium, foil, 50x50mm, thickness 0.25mm, annealed, 99.2%
Zr
GF83603909
Zirconium, foil, 50x50mm, thickness 0.5mm, annealed, 99.2%
Zr
GF04051886
Zirconium, foil, 50x50mm, thickness 2.0mm, annealed, 99.2%
Zr
GF14815593
Zirconium, foil, 10mm disks, thickness 0.0125mm, 99.2%
Zr
GF58113019
Zirconium, foil, 10mm disks, thickness 0.015mm, 99.2%
Zr
GF67870468
Zirconium, foil, 10mm disks, thickness 0.016mm, 99.2%
Zr
GF58442070
Zirconium, foil, 10mm disks, thickness 0.01mm, 99.2%
Zr
GF54823642
Zirconium, foil, 10mm disks, thickness 0.01mm, 99.2%
Zr
GF75562869
Zirconium, foil, 10mm disks, thickness 0.025mm, annealed, 99.2%
Zr
GF55287221
Zirconium, foil, 10mm disks, thickness 0.02mm, 99.2%
Zr
GF38009044
Zirconium, foil, 10mm disks, thickness 0.05mm, annealed, 99.2%
Zr
GF89139594
Zirconium, foil, 10mm disks, thickness 0.075mm, annealed, 99.2%
Zr
GF22007595
Zirconium, foil, 10mm disks, thickness 0.125mm, annealed, 99.2%
Zr