EMAIL THIS PAGE TO A FRIEND
Nanoscale research letters
A composite CdS thin film/TiO2 nanotube structure by ultrafast successive electrochemical deposition toward photovoltaic application.
PMID 25520588
Abstract
Fabricating functional compounds on substrates with complicated morphology has been an important topic in material science and technology, which remains a challenging issue to simultaneously achieve a high growth rate for a complex nanostructure with simple controlling factors. Here, we present a novel simple and successive method based on chemical reactions in an open reaction system manipulated by an electric field. A uniform CdS/TiO2 composite tubular structure has been fabricated in highly ordered TiO2 nanotube arrays in a very short time period (~90xa0s) under room temperature (RT). The content of CdS in the resultant and its crystalline structure was tuned by the form and magnitude of external voltage. The as-formed structure has shown a quite broad and bulk-like light absorption spectrum with the absorption of photon energy even below that of the bulk CdS. The as-fabricated-sensitized solar cell based on this composite structure has achieved an efficiency of 1.43% without any chemical doping or co-sensitizing, 210% higher than quantum dot-sensitized solar cell (QDSSC) under a similar condition. Hopefully, this method can also easily grow nanostructures based on a wide range of compound materials for energy science and electronic technologies, especially for fast-deploying devices.