EMAIL THIS PAGE TO A FRIEND
Journal of gastroenterology and hepatology
Effect of rebamipide on gastric bleeding and ulcerogenic responses induced by aspirin plus clopidogrel under stimulation of acid secretion in rats.
PMID 25521732
Abstract
We examined the prophylactic effect of rebamipide on gastric bleeding induced by the perfusion of aspirin (acetylsalicylic acid [ASA]) plus clopidogrel under the stimulation of acid secretion in rats. Under urethane anesthesia, acid secretion was stimulated by the i.v. infusion of histamine (8 mg/kg/h), and the stomach was perfused with 25 mmol/L ASA at a rate of 0.4 mL/min. Gastric bleeding was evaluated as the concentration of hemoglobin in the perfusate. Clopidogrel (30 mg/kg) was given p.o. 24 h before the perfusion. Rebamipide (3-30 mg/kg) or other antiulcer drugs were given i.d. before the ASA perfusion. Slight gastric bleeding or damage was observed with the perfusion of ASA under the stimulation of acid secretion, whereas these responses were significantly increased in the presence of clopidogrel. Both omeprazole and famotidine inhibited acid secretion and prevented these responses to ASA plus clopidogrel. Rebamipide had no effect on acid secretion, but dose-dependently prevented gastric bleeding in response to ASA plus clopidogrel, with the degree of inhibition being almost equivalent to that of the antisecretory drugs, and the same effects were obtained with the gastroprotective drugs, irsogladine and teprenone. These agents also reduced the severity of gastric lesions, although the effects were less than those of the antisecretory drugs. These results suggest that the antiplatelet drug, clopidogrel, increases gastric bleeding induced by ASA under the stimulation of acid secretion, and the gastroprotective drug, rebamipide, is effective in preventing the gastric bleeding induced under such conditions, similar to antisecretory drugs.