PloS one
A recombinant avian leukosis virus subgroup j for directly monitoring viral infection and the selection of neutralizing antibodies.
PMID 25522008
Abstract
Avian leukosis virus subgroup J (ALV-J) has induced serious clinical outbreaks and has become a serious infectious disease of chickens in China. We describe here the creation of a recombinant ALV-J tagged with the enhanced green fluorescent protein (named rHPRS-103EGFP). We successfully utilize the rHPRS-103EGFP to visualize viral infection and for development of a simplified serum-neutralization test.