EMAIL THIS PAGE TO A FRIEND
Natural product communications
Essential oil from the heartwood of Taiwan fir ameliorates LPS-induced inflammatory response by inhibiting the activation of mitogen-activated protein kinase.
PMID 25522551
Abstract
The essential oil from the heartwood of Taiwan fir (EOTC) was demonstrated to exhibit anti-inflammatory activity in lipopolysaccharide (LPS)-activated mouse macrophages. EOTC reduced nitrite oxide levels and inducible nitrite oxide synthase expression in, and tumor necrosis factor-α and interleukin-6 secretion by, LPS-activated macrophages without affecting cyclooxygenase-2 expression. EOTC reduced the levels of interleukin-lβ precursor induced by LPS and decreased the NLRP3 inflammasome-derived interleukin-lβ secretion induced by LPS and adenosine triphosphate. In addition, the phosphorylation levels of ERKI/2, JNK1/2, and p38 in LPS-activated macrophages were reduced by EOTC. Furthermore, EOTC was composed of oxygenated sesquiterpenes (68.4%), sesquiterpene hydrocarbons (28.9%) and diterpenes (0.9%). The major compounds of the oxygenated sesquiterpenes were τ-cadinol (23.9%), α-cadinol (21.1%) and cedrol (16.9%). These findings suggest that EOTC may be a candidate for the development of anti-inflammatory agents for preventing and ameliorating inflammation-related diseases.