EMAIL THIS PAGE TO A FRIEND
Stroke
MicroRNA-424 protects against focal cerebral ischemia and reperfusion injury in mice by suppressing oxidative stress.
PMID 25523055
Abstract
We previously showed that the microRNA miR-424 protects against permanent cerebral ischemic injury in mice by suppressing microglia activation. This study investigated the role of miR-424 in transient cerebral ischemia in mice with a focus on oxidative stress-induced neuronal injury. Transient cerebral ischemia was induced in C57/BL6 mice by middle cerebral artery occlusion for 1 hour followed by reperfusion (ischemia/reperfusion). The miR-424 level in the peri-infarct cortex was quantified. Mice were also administered miR-424 angomir by intracerebroventricular injection. Cerebral infarct volume, neuronal apoptosis, and levels of oxidative stress markers and antioxidants were evaluated. In an in vitro experiment, primary cortical neurons were exposed to H2O2 and treated with miR-424 angomir, nuclear factor erythroid 2-related factor 2 siRNA, and superoxide dismutase (SOD) inhibitor; cell activity, lactate dehydrogenase release, malondialdehyde level, and manganese (Mn)SOD activity were then evaluated. MiR-424 levels in the peri-infarct cortex increased at 1 and 4 hours then decreased 24 hours after reperfusion. Treatment with miR-424 decreased infarct volume and inhibited neuronal apoptosis after ischemia/reperfusion, reduced reactive oxygen species and malondialdehyde levels in the cortex, and increased the expression and activation of MnSOD as well as the expression of extracellular SOD and the redox-sensitive transcription factor nuclear factor erythroid 2-related factor. In neuronal cultures, miR-424 treatment abrogated H2O2-induced injury, as evidenced by decreased lactate dehydrogenase leakage and malondialdehyde level and increased cell viability and MnSOD activity; the protective effects of miR-424 against oxidative stress were reversed by nuclear factor erythroid 2-related factor knockdown and SOD inhibitor treatment. MiR-424 protects against transient cerebral ischemia/reperfusion injury by inhibiting oxidative stress.