EMAIL THIS PAGE TO A FRIEND
Analytical and bioanalytical chemistry
Biomarkers probed in saliva by surface plasmon resonance imaging coupled to matrix-assisted laser desorption/ionization mass spectrometry in array format.
PMID 25524230
Abstract
Detection of protein biomarkers is of major interest in proteomics. This work reports the analysis of protein biomarkers directly from a biological fluid, human saliva, by surface plasmon resonance imaging coupled to mass spectrometry (SPRi-MS), using a functionalized biochip in an array format enabling multiplex SPR-MS analysis. The SPR biochip presented a gold surface functionalized by a self-assembled monolayer of short poly(ethylene oxide) chains carrying an N-hydroxysuccinimide end-group for the immobilization of antibodies. The experiments were accomplished without any sample pre-purification or spiking with the targeted biomarkers. SPRi monitoring of the interactions, immune capture from the biochip surface, and finally on-chip matrix-assisted laser desorption/ionization-MS structural identification of two protein biomarkers, salivary α-amylase and lysozyme, were successively achieved directly from saliva at the femtomole level. For lysozyme, the on-chip MS identification was completed by a proteomic analysis based on an on-chip proteolysis procedure and a peptide mass fingerprint.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
83275
4-Pyrrolidinopyridine, purum, ≥98.0% (NT)
C9H12N2
213373
4-Pyrrolidinopyridine, 98%
C9H12N2
242381
Benzoic acid, ACS reagent, ≥99.5%
C7H6O2
33045
Benzoic acid, for calorimetrical determination (approx. 26460 J/g)
C7H6O2
06185
Benzoic acid, Standard for quantitative NMR, TraceCERT®
C7H6O2
1055002
Benzoic acid, United States Pharmacopeia (USP) Reference Standard
C7H6O2
33047
Benzoic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9% (alkalimetric)
C7H6O2
18102
Benzoic acid, meets analytical specification of Ph. Eur., BP, USP, FCC, E210, 99.5-100.5% (alkalimetric)
C7H6O2
12356
Benzoic acid, tested according to Ph.Eur.
C7H6O2
PHR1050
Benzoic acid, Pharmaceutical Secondary Standard; Certified Reference Material
C7H6O2
12353
Benzoic acid, certified reference material for titrimetry, certified by BAM according to ISO 17025, ≥99.5%
C7H6O2
47849
Benzoic acid, analytical standard
C7H6O2
109479
Benzoic acid, ReagentPlus®, 99%
C7H6O2
W213101
Benzoic acid, ≥99.5%, FCC, FG
C7H6O2
W213128
Benzoic acid, natural, ≥99.5%, FCC, FG
C7H6O2
Y0001470
Benzoic acid, European Pharmacopoeia (EP) Reference Standard
C7H6O2
491861
Benzoic acid-12C7, 99.9 atom % 12C
12C7H6O2
23128
L-Lysine, analytical standard
C6H14N2O2
62850
L-Lysine solution, purum, 50% in H2O
C6H14N2O2
76170
Melting point standard 121-123°C, analytical standard
C7H6O2
73983
Mettler-Toledo Calibration substance ME 18555, Benzoic acid, analytical standard, for the calibration of the thermosystem 900, traceable to primary standards (LGC)
C7H6O2
8S61336
SS Benzoic Acid, 2000 μg/mL in dichloromethane, analytical standard
C7H6O2