EMAIL THIS PAGE TO A FRIEND
The Journal of thoracic and cardiovascular surgery
What the Human Genome Project hasn't told us: the epigenetics of development of esophageal squamous cell cancer.
PMID 25524691