EMAIL THIS PAGE TO A FRIEND
Journal of neuroscience research
Different protective and reparative effects of olmesartan in stroke according to time of administration and withdrawal.
PMID 25524827
Abstract
Angiotensin type 1 receptor blockers (ARBs) have induced improved functional recovery and reduced infarct volume in experimental animal models of stroke. Clinical data have indicated a positive correlation between prestroke treatment with ARBs and reduced stroke severity and better outcomes; however, the mechanisms of these beneficial effects are not yet well understood. This study compares the protective and possible reparative effects of continuous oral treatment with olmesartan (OLM) with OLM pretreatment and withdrawal after permanent middle cerebral artery occlusion (pMCAO) in rats. Fifty-two Sprague-Dawley rats were randomly assigned to five groups: MCAO(-/OLM) (OLM 10 mg/kg/day for 14 days after infarct), MCAO(OLM/OLM) (OLM 10 mg/kg/day for 7 days before and 14 days after infarct), MCAO(OLM/-) (OLM 10 mg/kg/day for 7 days before infarct), sham, and control. We analyzed functional recovery; lesion size; cell death; expression of the pro-oxidant enzyme NADPH oxidase 4 (NOX-4); isolectin-B4; and repair markers such as glial fibrillary acidic protein, vascular endothelial growth factor (VEGF), and brain-derived neurotrophic factor (BDNF). All of the OLM-treated groups showed significantly better functional scores and reduced infarct sizes and cell death compared with the control group 14 days after pMCAO. Levels of NOX-4, VEGF, and BDNF were significantly lower in the brains of the MCAO(OLM/OLM) and sham groups compared with the other groups. OLM treatment improved functional recovery and reduced lesion size and cell death after cerebral ischemia. Only the continuous administration of OLM before and after stroke reduced oxidative stress levels, with better tissue preservation, without triggering brain repair marker activation.