EMAIL THIS PAGE TO A FRIEND
The Journal of infectious diseases
Host-pathogen Interaction at the Intestinal Mucosa Correlates With Zoonotic Potential of Streptococcus suis.
PMID 25525050
Abstract
Streptococcus suis has emerged as an important cause of bacterial meningitis in adults. The ingestion of undercooked pork is a risk factor for human S. suis serotype 2 (SS2) infection. Here we provide experimental evidence indicating that the gastrointestinal tract is an entry site of SS2 infection. We developed a noninvasive in vivo model to study oral SS2 infection in piglets. We compared in vitro interaction of S. suis with human and porcine intestinal epithelial cells (IEC). Two out of 15 piglets showed clinical symptoms compatible with S. suis infection 24-48 hours after ingestion of SS2. SS2 was detected in mesenteric lymph nodes of 40% of challenged piglets. SS2 strains isolated from patients showed significantly higher adhesion to human IEC compared to invasive strains isolated from pigs. In contrast, invasive SS9 strains showed significantly higher adhesion to porcine IEC. Translocation across human IEC, which occurred predominately via a paracellular route, was significantly associated with clonal complex 1, the predominant zoonotic genotype. Adhesion and translocation were dependent on capsular polysaccharide production. SS2 should be considered a food-borne pathogen. S. suis interaction with human and pig IEC correlates with S. suis serotype and genotype, which can explain the zoonotic potential of SS2.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA039844
Anti-BTNL2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4200489
Anti-Occludin (C-terminal) antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody
SAB1406198
Anti-OCLN antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA005933
Anti-OCLN antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB3500301
Anti-OCLN antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2103797
Anti-OCLN, (N-terminal) antibody produced in rabbit, affinity isolated antibody
WH0004950M1
Monoclonal Anti-OCLN antibody produced in mouse, clone 1G7, purified immunoglobulin, buffered aqueous solution
AMAB90893
Monoclonal Anti-OCLN antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1608, purified immunoglobulin, buffered aqueous glycerol solution
AMAB90890
Monoclonal Anti-OCLN antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1567, purified immunoglobulin, buffered aqueous glycerol solution
AMAB90889
Monoclonal Anti-OCLN antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1555, purified immunoglobulin, buffered aqueous glycerol solution