EMAIL THIS PAGE TO A FRIEND
Science (New York, N.Y.)
A designed supramolecular protein assembly with in vivo enzymatic activity.
PMID 25525249
Abstract
The generation of new enzymatic activities has mainly relied on repurposing the interiors of preexisting protein folds because of the challenge in designing functional, three-dimensional protein structures from first principles. Here we report an artificial metallo-β-lactamase, constructed via the self-assembly of a structurally and functionally unrelated, monomeric redox protein into a tetrameric assembly that possesses catalytic zinc sites in its interfaces. The designed metallo-β-lactamase is functional in the Escherichia coli periplasm and enables the bacteria to survive treatment with ampicillin. In vivo screening of libraries has yielded a variant that displays a catalytic proficiency [(k(cat)/K(m))/k(uncat)] for ampicillin hydrolysis of 2.3 × 10(6) and features the emergence of a highly mobile loop near the active site, a key component of natural β-lactamases to enable substrate interactions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
PHR1424
Ampicillin Sodium, Pharmaceutical Secondary Standard; Certified Reference Material
C16H18N3NaO4S
A9518
Ampicillin sodium salt
C16H18N3NaO4S
A0166
Ampicillin sodium salt, powder, BioReagent, suitable for cell culture
C16H18N3NaO4S
A8351
Ampicillin sodium salt, BioXtra, suitable for cell culture
C16H18N3NaO4S
A6140
Ampicillin trihydrate, 96.0-100.5% anhydrous basis (HPLC)
C16H19N3O4S · 3H2O
31591
Ampicillin trihydrate, VETRANAL, analytical standard
C16H19N3O4S · 3H2O
A1100000
Ampicillin trihydrate, European Pharmacopoeia (EP) Reference Standard
C16H19N3O4S · 3H2O
PHR1393
Ampicillin Trihydrate, Pharmaceutical Secondary Standard; Certified Reference Material
C16H19N3O4S · 3H2O
31653
Zinc, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.9%, granular
Zn
96454
Zinc, purum, powder
Zn
209988
Zinc, dust, <10 μm, ≥98%
Zn
349410
Zinc, foil, thickness 1.0 mm, 99.99% trace metals basis
Zn
349429
Zinc, foil, thickness 0.5 mm, 99.999%
Zn
267619
Zinc, foil, thickness 0.25 mm, 99.999% trace metals basis
Zn
267600
Zinc, foil, thickness 0.1 mm, 99.999%
Zn
356018
Zinc, foil, thickness 0.25 mm, 99.9% trace metals basis
Zn
565148
Zinc, granular, −30 mesh±100, 99%
Zn
266345
Zinc, granular, −10-+50 mesh, ≥99.8%, ACS reagent
Zn
243469
Zinc, granular, 20-30 mesh, ACS reagent, ≥99.8%
Zn
243477
Zinc, ACS reagent, −30-+100 mesh, ≥99.8%, granular
Zn
215503
Zinc, mossy, 99.99% trace metals basis
Zn
215481
Zinc, mossy, ≥99%
Zn
578002
Zinc, nanopowder, 40-60 nm avg. part. size, ≥99% trace metals basis
Zn
266353
Zinc, pieces, 2-14 mesh, 99.9% trace metals basis
Zn
324930
Zinc, powder, <150 μm, 99.995% trace metals basis
Zn
267635
Zinc, shot, 5 mm, 99.999% trace metals basis
Zn
402583
Zinc, shot, <12 mm, 99.99% trace metals basis
Zn
266361
Zinc, sticks, diam. 7-10 mm, 99.97% trace metals basis
Zn
267929
Zinc, wire, diam. 1.0 mm, 99.999% trace metals basis
Zn
355976
Zinc, wire, diam. 0.25 mm, 99.999%
Zn