EMAIL THIS PAGE TO A FRIEND
Atherosclerosis
Dietary predictors of arterial stiffness in a cohort with type 1 and type 2 diabetes.
PMID 25528424
Abstract
To determine the dietary predictors of central blood pressure, augmentation index and pulse wave velocity (PWV) in subjects with type 1 and type 2 diabetes. Participants were diagnosed with type 1 or type 2 diabetes and had PWV and/or pulse wave analysis performed. Dietary intake was measured using the Dietary Questionnaire for Epidemiological Studies Version 2 Food Frequency Questionnaire. Serum lipid species and carotenoids were measured, using liquid chromatography electrospray ionization-tandem mass spectrometry and high performance liquid chromatography, as biomarkers of dairy and vegetable intake, respectively. Associations were determined using linear regression adjusted for potential confounders. PWV (nxa0=xa095) was inversely associated with reduced fat dairy intake (βxa0=xa0-0.01; 95% CIxa0-0.02,xa0-0.01; pxa0=xa00xa0