EMAIL THIS PAGE TO A FRIEND
Nature communications
Critical role of histone demethylase Jmjd3 in the regulation of CD4+ T-cell differentiation.
PMID 25531312
Abstract
Epigenetic factors have been implicated in the regulation of CD4(+) T-cell differentiation. Jmjd3 plays a role in many biological processes, but its in vivo function in T-cell differentiation remains unknown. Here we report that Jmjd3 ablation promotes CD4(+) T-cell differentiation into Th2 and Th17 cells in the small intestine and colon, and inhibits T-cell differentiation into Th1 cells under different cytokine-polarizing conditions and in a Th1-dependent colitis model. Jmjd3 deficiency also restrains the plasticity of the conversion of Th2, Th17 or Treg cells to Th1 cells. The skewing of T-cell differentiation is concomitant with changes in the expression of key transcription factors and cytokines. H3K27me3 and H3K4me3 levels in Jmjd3-deficient cells are correlated with altered gene expression through interactions with specific transcription factors. Our results identify Jmjd3 as an epigenetic factor in T-cell differentiation via changes in histone methylation and target gene expression.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
07-452
Anti-dimethyl-Histone H3 (Lys27) Antibody, 200 µg, Use Anti-dimethyl-Histone H3 (Lys27) Antibody (rabbit polyclonal antibody) validated in Mplex, PIA, WB, ChIP-seq, DB to detect dimethyl-Histone H3 (Lys27) also known as H3K27me2, Histone H3 (di methyl K27).
07-030
Anti-dimethyl-Histone H3 (Lys4) Antibody, 200 µL, Use Anti-dimethyl-Histone H3 (Lys4) Antibody (rabbit polyclonal antibody) is published and validated in ChIP, DB, ICC, IF, WB, PIA, ChIP-seq. to detect dimethyl-Histone H3 (Lys4) also known as H3K4me2, Histone H3 (di methyl K4).
07-441
Anti-dimethyl-Histone H3 (Lys9) Antibody, 100 µg, Use Anti-dimethyl-Histone H3 (Lys9) Antibody (Rabbit Polyclonal Antibody) validated in DB, ICC, PIA, WB to detect dimethyl-Histone H3 (Lys9) also known as H3K9me2, Histone H3 (di methyl K9).
07-448
Anti-monomethyl-Histone H3 (Lys27) Antibody, 200 µg, Anti-monomethyl-Histone H3 (Lys27) Antibody is a rabbit polyclonal antibody to detect Histone H3 monomethylated at lysine 27. This purified Ab, also known as Anti-H3K27me1 is specificity verified by dot blot (DB), published in peer reviewed journals and validated in ICC, WB, ChIP, ChIP-seq.
07-436
Anti-monomethyl-Histone H3 (Lys4) Antibody, 200 µg, Anti-monomethyl-Histone H3 (Lys4) Antibody is a rabbit polyclonal antibody for detection of monomethyl-Histone H3 (Lys4) also known as H3K4me1, Histone H3 (mono methyl K4) & has been validated in ICC, IF, PIA, WB, ChIP-seq, DB.
07-450
Anti-monomethyl-Histone H3 (Lys9) Antibody, 100 µg, Anti-monomethyl-Histone H3 (Lys9) Antibody is a Rabbit Polyclonal Antibody for detection of Histone H3 monomethylated on lysine 9 (H3K9me1). This purified antibody is specificity verified by dot blot (DB), published in peer reviewed journals, and has been validated in ICC, PIA & WB.
07-449
Anti-trimethyl-Histone H3 (Lys27) Antibody, 200 µg, Anti-trimethyl-Histone H3 (Lys27), also known as Anti-H3K27me3, is a highly published Rabbit Polyclonal Antibody. This protein A purified antibody is dot blot tested for trimethylated lysine 27 specificity and validated in WB, ChIP, ChIP-seq, ICC, IP.
07-473
Anti-trimethyl-Histone H3 (Lys4) Antibody, 100 µL, Anti-trimethyl-Histone H3 (Lys4) Antibody is a rabbit polyclonal antibody for detection of Histone H3 trimethylated at lysine 4. Also known as Anti-H3K4me3, this highly specific and well published antibody has been validated in ChIP, DB, WB, PIA, ChIP-seq.
07-442
Anti-trimethyl-Histone H3 (Lys9) Antibody, 100 µg, Use Anti-trimethyl-Histone H3 (Lys9) Antibody (rabbit polyclonal antibody) validated in DB, ICC, Mplex, PIA, WB, ChIP-seq to detect trimethyl-Histone H3 (Lys9) also known as H3K9me3, Histone H3 (tri methyl K9).